Voriconazole ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: (2R,3S)-2-(2,4-Difluorophenyl)-3-(5-fluoropyrimidin-4-yl)-1-(1H-1,2,4-triazol-1-yl)butan-2-ol, (2R,3S)-2-(2,4-Difluorophenyl)-3-(5-fluoro-4-pyrimidinyl)-1-(1H-1,2,4-triazol-1-yl)-2-butanol

V0116-100MG V0116-1G
TCIAV0116-100MGEA 30.8 EUR
TCIAV0116-100MG TCIAV0116-1G
Voriconazole ≥98.0% (by HPLC)
Voriconazole
Formule: C₁₆H₁₄F₃N₅O
MW: 349.32 g/mol
MDL Nummer: MFCD00905717
CAS nummer: 137234-62-9
UN: 2811
ADR: 6.1 III
Merck Index: 14,10033
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a884f2bff6fb1b1c59b58f26f22a18b399cdc96e&vid=a884f2bff6fb1b1c59b58f26f22a18b399cdc96e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 130.0 to 134.0 deg-C
Specific rotation [a]20/D -58.0 to -62.0 deg (C=1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR