4-Vinylpyridine ≥95.0% (by GC) stabilised

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar


Stabilisatie: Stabilised with hydroquinone (HQ)

V0150-25ML V0150-500ML
TCIAV0150-25MLEA 42.9 EUR
TCIAV0150-25ML TCIAV0150-500ML
4-Vinylpyridine ≥95.0% (by GC) stabilised
4-Vinylpyridine
Formule: C₇H₇N
Kookpunt: 59°C
Vlampunt: 56°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00006447
CAS nummer: 100-43-6
EINECS: 202-852-0
UN: 3073
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af12d38eddb30b6b61910f7bcc1bb59991b70232&vid=44d265207916ee1ded50e73b653ba848e6ba936c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR