Vanillyl butyl ether ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 4-(Butoxymethyl)-2-methoxyphenol

V0161-25G V0161-5G
TCIAV0161-25GEA 67.1 EUR
TCIAV0161-25G TCIAV0161-5G
Vanillyl butyl ether ≥98.0% (by GC)
Vanillyl butyl ether
Formule: C₁₂H₁₈O₃
Kookpunt: 241°C
MDL Nummer: MFCD00238529
CAS nummer: 82654-98-6

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR