Xanthane hydride ≥95.0% (by HPLC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
5-Amino-3H-1,2,4-dithiazole-3-thione
X0001-25G
X0001-500G
X0001-100G
TCIAX0001-25GEA
67.1
EUR
TCIAX0001-25G
TCIAX0001-500G
TCIAX0001-100G
Xanthane hydride ≥95.0% (by HPLC)
Xanthane hydride
Formule:
C₂H₂N₂S₃ |
MDL Nummer:
MFCD00051660 CAS nummer: 6846-35-1 EINECS: 229-933-3 |
|
Specification Test Results
Appearance | Light yellow to Brown powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Solubility in H2SO4 | transparency |
Water | max. 1.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...