Xipamide ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

X0079-250MG X0079-50MG
TCIAX0079-250MGEA 165 EUR
TCIAX0079-250MG TCIAX0079-50MG
Xipamide ≥98.0% (by HPLC)
Xipamide
Formule: C₁₅H₁₅ClN₂O₄S
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00865927
CAS nummer: 14293-44-8
EINECS: 238-216-4
Merck Index: 14,10079

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR