Zoledronic acid monohydrate ≥98.0% (by HPLC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
1-Hydroxy-2-(1-imidazolyl)ethane-1,1-diphosphonic acid monohydrate, Reclast, Zoledronate monohydrate, ZOL 446 monohydrate, NDC-zoledronate monohydrate, CGP42446A monohydrate, (1-Hydroxy-2-(1H-imidazol-1-yl)ethane-1,1-diyl)diphosphonic acid monohydrate, (1-Hydroxy-2-(1H-imidazol-1-yl)ethane-1,1-diyl)diphosphonic acid hydrate, Zometa
Z0031-1G
Z0031-5G
TCIAZ0031-1GEA
51.7
EUR
TCIAZ0031-1G
TCIAZ0031-5G
Zoledronic acid monohydrate ≥98.0% (by HPLC, titration analysis)
Zoledronic acid monohydrate
Formule:
C₅H₁₂N₂O₈P₂ Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00867791 CAS nummer: 165800-06-6 Merck Index: 14,10187 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % (calcd.on anh.substance) |
Water | 5.5 to 8.8 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...