Industrial gloves, HyFlex® 11-751
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11-751/6 11-751/7 11-751/8 11-751/9 11-751/10 11-751/11
ANSE11-751/6EA 207 EUR
ANSE11-751/6 ANSE11-751/7 ANSE11-751/8 ANSE11-751/9 ANSE11-751/10 ANSE11-751/11
Industrial gloves, HyFlex® 11-751
Handschoenen
The HyFlex® 11-751 glove features a 10G lightweight, medium-duty liner with INTERCEPT TECHNOLOGY® yarns that provide EN ISO Cut Level C protection for high levels of cut resistance.
  • Silicone-free
  • Increased worker confidence when handling small, sharp parts
  • Exceptional abrasion resistance
The comfortable and flexible plaited liner also features an optimized Lycra® content that is soft and cool on the skin for all-day comfort.
The exceptional precision, dexterity, and flexibility when handling small, sharp parts increases worker confidence while on the job. Durable and launderable with minimal shrinkage and excellent recovery after laundering, extending glove life.
INTERCEPT TECHNOLOGY® yarn has been tested to perform at or exceed industry standards. This proprietary technology offers best-in-class cut protection and increased comfort and dexterity. The palm coating design improves worker grip, extends glove life in abrasive conditions and allows the back of hand to breathe.
For use in the aerospace, automotive, machinery and equipment, and metal fabrication industries for assembly of small sharp parts, handling small sharp metal parts, light-duty stamping and maintenance.
Certificaten: EN 388:2016 (4X43C); Category II
Verpakking: 12 pairs per bag.