Industrial gloves, HyFlex® 11-931
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11-931/10 11-931/6 11-931/8 11-931/11 11-931/7 11-931/9
ANSE11-931/10EA 216 EUR
ANSE11-931/10 ANSE11-931/6 ANSE11-931/8 ANSE11-931/11 ANSE11-931/7 ANSE11-931/9
Industrial gloves, HyFlex® 11-931
Handschoenen
The lightest weight cut resistant and oil repellent glove providing enhanced worker comfort and safety, with strategically reinforced 'high wear' regions for extended-use life.
  • RIPEL™ technology prevents liquids and oil from making skin contact keeping hands safe and dry
  • Thumb crotch reinforcement technology for up to 12x the durability of the inherently weak thumb crotch region*
  • ANSELL GRIP™ technology delivers superior grip performance for safer handling of wet or oily parts
  • FORTIX™ technology provides extended glove life and high abrasion resistance
More Product Information
Order Now
Specificaties
Model
Ambidexter/handspecifiek Handspecifiek Handspecifiek
Environmentally Preferable
Voor eenmalig gebruik/herbruikbaar Herbruikbaar Herbruikbaar
Latex/latexvrij Latexvrij Latexvrij
Gepoederd/poedervrij poedervrij poedervrij
Voeringmateriaal Dyneema® Dyneema®
Afwerking Foam nitrile palm coating Schuim nitril palm coating
Steken per inch (Gauge) 18 18
Manchetstijl Gebreide manchet Gebreide manchet
EN-norm EN 388:2016 (4X42B) EN 388:2016 (4X42B)
Antistatisch + +