Industrial gloves, HyFlex® 11-937
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11-937/6 11-937/8 11-937/11 11-937/10 11-937/9 11-937/7
ANSE11-937/6EA 221 EUR
ANSE11-937/6 ANSE11-937/8 ANSE11-937/11 ANSE11-937/10 ANSE11-937/9 ANSE11-937/7
Industrial gloves, HyFlex® 11-937
Handschoenen
The lightest weight cut resistant and oil repellent glove providing enhanced worker comfort and safety, with strategically reinforced 'high wear' regions for extended-use life.
  • RIPEL™ technology prevents liquids and oil from making skin contact keeping hands safe and dry
  • Thumb crotch reinforcement technology for up to 12x the durability of the inherently weak thumb crotch region*
  • ANSELL GRIP™ technology delivers superior grip performance for safer handling of wet or oily parts
  • FORTIX™ technology provides extended glove life and high abrasion resistance
  • ZONZ™ comfort fit technology provides dexterity and comfort
The 18 gauge liner featuring Dyneema® Diamond Technology provides high dexterity and comfort. The foam nitrile coating and integrated thumb crotch reinforcement (TCR) offer long-lasting protection and 12x better wear life than unreinforced gloves based on field tests. Foam nitrile ¾ coating provides liquid repellence. A silicone free design insures full product protection.
Recommended for: Inspection, selecting, checking parts; assembly & inspection of components; fastening components; chassis assembly, body trimming; adjusting parts; delivery of parts to production line; equipment repair & maintenance; engine transmission and repair; oil, fluids and filter change; inspection (picking, checking and marking); feeding the press and removing spare parts from it; adjusting panels parts components; press shop operations.
*compared to unreinforced TCR competitor gloves.
Certificaten: EN 388:2016 (4X42B); Cat. II
Oeko-Tex; REACH compliant.
Verpakking: 12 pairs per bag.