5-Aminoisophthalic acid 98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
181331000
181330250
ACRO181331000EA
63
EUR
ACRO181331000
ACRO181330250
5-Aminoisophthalic acid 98%
5-Aminoisophthalic acid
Formule:
C₈H₇NO₄ Kookpunt: 515 °C (1013 hPa) Smeltpunt: >300 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00002522 CAS nummer: 99-31-0 EINECS: 202-748-5 |
Specification Test Results
Appearance | White to beige to light brown Powder |
Infrared spectrum | Conforms |
HPLC | ≥94 % |
Specific optical rotation | -143° to -153° (20°C, 589 nm) (1% in ethanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...