Benzeneselenenyl bromide 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
223060050 223060250
ACRO223060050EA 70.7 EUR
ACRO223060050 ACRO223060250
Benzeneselenenyl bromide 98%
Benzeneselenenyl bromide
Formule: (C₆H₅)SeBr
Kookpunt: 107…108 °C (15 mmHg)
Smeltpunt: 60…62 °C
Dichtheid: 1,898 g/cm³ (65.5 °C)
MDL Nummer: MFCD00000047
CAS nummer: 34837-55-3
EINECS: 252-238-1
UN: 2928
ADR: 6.1,II
Order Now
Specification Test Results
Appearance Dark brown to black Crystals
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %