Wipe rolls, WypAll® L10 compact food and hygiene
Leverancier: KIMBERLY CLARK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7225 7236
KIMB7225EA 217 EUR
KIMB7225 KIMB7236
Wipe rolls, WypAll® L10 compact food and hygiene
Schoonmaakdoekjes Doekjes en handdoeken voor eenmalig gebruik Doekjes voor algemeen gebruik
Compact disposable paper wipes are 1-ply and designed for light-duty wiping in areas such as food preparation and service surfaces.
  • Compact roll design is small, portable, and easy to handle
  • Ease of use helps to support hygiene standards and compliance while also saving space
  • WypAll® wiping paper is designed to be used once with detergent or disinfectant
These dry cleaning wipes have been tested as compatible for use in spray and wipe application with peracetic acid, hydrogen peroxide, and quats (quaternary ammonium compounds). Spray and wipingMore Product Information