4-Iodo-o-toluidine
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: 2-Amino-5-iodotoluene, 4-Iodo-2-methylaniline
RDP00085.10 RDP00085.50
MAYBRDP00085.10EA 0 EUR
MAYBRDP00085.10 MAYBRDP00085.50
4-Iodo-o-toluidine
4-Iodo-o-toluidine
Formule: C₇H₈IN
Kookpunt: 279 °C (1013 hPa)
Smeltpunt: 91…92 °C
MDL Nummer: MFCD00025299
CAS nummer: 13194-68-8
EINECS: 236-154-2
UN: 2811
ADR: 6.1,III