Chloramine-T (sodium salt) trihydrate 98%
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Purity: 98%
CAS No.: 7080-50-4
Molecular Formula: C7H7ClNNaO2S.3H2O
Formula Weight: 281.69 (227.67anhy)
Flash Point: 192°(377°F)
Physical Form: Crystalline
Sensitivity: Moisture Sensitive
Appearance: White to pale yellow
Odour: Chlorine-like
pH: 8-10
Melting Point: 167-170°
Beilstein Reference: 3924168
Merck Reference: 14,2075
UN No.: 3263
MDL No.: MFCD00149066
Formule:
CH₃C₆H₄SO₂NClNa·3H₂O Smeltpunt: 167…170 °C Dichtheid: 1,43 g/cm³ (20 °C) Vlampunt: 192 °C (377 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00149066 CAS nummer: 7080-50-4 EINECS: 204-854-7 UN: 3263 ADR: 8,III Merck Index: 14,2075 |