1,4-Cyclohexanediamine (cis and trans mixture) cis- and trans-mixture ≥97.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: Cyclohexane-1,4-diamine, 1,4-Diaminocyclohexane

C0814-25ML C0814-5ML
TCIAC0814-25MLEA 125 EUR
TCIAC0814-25ML TCIAC0814-5ML
1,4-Cyclohexanediamine (cis and trans mixture) cis- and trans-mixture ≥97.0% (by GC, titration analysis)
1,4-Cyclohexanediamine (cis and trans mixture)
Formule: C₆H₁₄N₂
Kookpunt: 199 °C (1013 hPa)
Smeltpunt: 70 °C
MDL Nummer: MFCD00035862
CAS nummer: 3114-70-3
EINECS: 221-483-6
UN: 3259
ADR: 8,III

Order Now

Specification Test Results

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 97.0 % (total of isomers)
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR