Cyclopropyl methyl ketone 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: 1-Cyclopropylethanone
111710250 111711000 111715000
ACRO111710250EA 30.3 EUR
ACRO111710250 ACRO111711000 ACRO111715000
Cyclopropyl methyl ketone 98%
Cyclopropyl methyl ketone
Formule: C₅H₈O
Kookpunt: 111 °C (760 mmHg)
Smeltpunt: –68 °C
Dichtheid: 0,898 g/cm³
Vlampunt: 21 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001297
CAS nummer: 765-43-5
EINECS: 212-146-4
UN: 2924
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4220 to 1.4260 (20°C, 589 nm)
Water ≤0.2 %
GC ≥97.5 %