Diphenylcyclopropenone for synthesis, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 2,3-Diphenyl-2-cyclopropen-1-one, 2,3-Diphenylcycloprop-2-en-1-one, 2,3-Diphenyl-2-cyclopropen-1-on, 1,2-Diphenylcyclopropen-3-one
M-Clarity™ quality level = MQ200
8.20533.0001 8.20533.0005 8.20533.8500
8.20533.0001EA 110 EUR
8.20533.0001 8.20533.0005 8.20533.8500
Diphenylcyclopropenone for synthesis, Sigma-Aldrich®
Diphenylcyclopropenone
Formule: C₁₅H₁₀O
MW: 206.24 g/mol
Kookpunt: 407
Smeltpunt: 112…113 °C
Dichtheid: 1.1 g/cm³ (20 °C)
MDL Nummer: MFCD00001311
CAS nummer: 886-38-4
EINECS: 212-948-4
Merck Index: 12,03366
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Melting Point 118 - 122 °C
Molar Mass 206.24 g/mol
Hill Formula C15H10O