1-(4-Chlorophenyl)piperazine dihydrochloride
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AC10992.1 AC10992.10 AC10992.50
MAYBAC10992.1EA 0 EUR
MAYBAC10992.1 MAYBAC10992.10 MAYBAC10992.50
1-(4-Chlorophenyl)piperazine dihydrochloride
1-(4-Chlorophenyl)piperazine dihydrochloride
Formule: C₁₀H₁₅Cl₃N₂
MDL Nummer: MFCD00012766
CAS nummer: 38869-46-4
EINECS: 254-165-0