4-Aminoantipyrine analytical reagent, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 4-Amino-2,3-dimethyl-1-phenyl-3-pyrazolin-5-one, Ampyrone, 4-Aminophenazone
M-Clarity™ quality level = MQ100
1.07293.0010 1.07293.0100
1.07293.0010EA 72.7 EUR
1.07293.0010 1.07293.0100
4-Aminoantipyrine analytical reagent, Supelco®
4-Aminoantipyrine
Formule: C₁₁H₁₃N₃O
MW: 203.24 g/mol
Smeltpunt: 107…109 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003145
CAS nummer: 83-07-8
EINECS: 201-452-3
Merck Index: 13,00596
Order Now
Specification Test Results
Solubility 56 g/l (20 °C)
Melting Point 107 - 109 °C
Molar Mass 203.24 g/mol
Bulk Density 430 kg/m3
pH 7.1 (100 g/l, H2O, 20 °C) (slurry)