Industrial gloves, HyFlex® 11-528
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11-528/9 11-528/6 11-528/7 11-528/8 11-528/10 11-528/11
ANSE11-528/9EA 156 EUR
ANSE11-528/9 ANSE11-528/6 ANSE11-528/7 ANSE11-528/8 ANSE11-528/10 ANSE11-528/11
Industrial gloves, HyFlex® 11-528
Handschoenen
This glass-free INTERCEPT liner provides increased cut protection and lasting comfort and protection for hard-working hands.
  • Skin friendly for increased comfort
  • INTERCEPT Technology® for enhanced cut protection
  • Enhanced flexibility for range of motion and tactility for handling small parts
  • Grip performance for safe handling in dry to light oily applications
  • NBR foam coating provides high grip and abrasion (ANSI 4)
  • DMF-free
Ideal for use in the automotive and transportation industry.
Recommended for: Handling metal sheets & panels; handlingMore Product Information
Order Now
Specificaties
Model
Ambidexter/handspecifiek Handspecifiek Handspecifiek
Environmentally Preferable
Voor eenmalig gebruik/herbruikbaar Herbruikbaar Herbruikbaar
Voeringmateriaal HPPE/basalt/polyester/Spandex HPPE/basalt/polyester/Spandex
Afwerking Palm nitrile coated
Steken per inch (Gauge) 18 18
Manchetstijl Gebreide manchet Gebreide manchet
EN-norm EN 388:2016 (3X32B) EN 388:2016 (3X32B)
Antistatisch + +
Goedgekeurd voor voedselcontact + +