Industrial gloves, HyFlex® 11-135
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11-135/10 11-135/6 11-135/7 11-135/8 11-135/9
ANSE11-135/10EA 63.2 EUR
ANSE11-135/10 ANSE11-135/6 ANSE11-135/7 ANSE11-135/8 ANSE11-135/9
Industrial gloves, HyFlex® 11-135
Handschoenen
Glove with excellent comfort and dexterity. Knitted with a seamless non-stop process using high-tenacity polyamid continuous thread.
  • Antistatic properties of the glove allow the user to be protected against electrostatic discharge
  • Fine and stretchable stitches for an excellent comfort
  • Stretchable PU coating
  • Excellent comfort and dexterity
  • Low linting
Recommended industries: Automotive, electronics, machinery and equipment, Life Sciences
Certificaten: EN 388:2016 (113XA); Cat. II
Verpakking: 12 pairs per bag.
Order Now
Specificaties
Model 11-135 11-135
Ambidexter/handspecifiek Handspecifiek Handspecifiek
Environmentally Preferable
Voor eenmalig gebruik/herbruikbaar Herbruikbaar Herbruikbaar
Latex/latexvrij Latex Latex
Voeringmateriaal Polyamide / koper Polyamide / koper
Afwerking Fingertips PU coated
Steken per inch (Gauge) 13 13
Manchetstijl Gebreide manchet Gebreide manchet
EN-norm EN 388:2016 (113XA) EN 388:2016 (113XA)
Antistatisch + +