Industrial gloves, HyFlex® 11-515
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11-515/11 11-515/6 11-515/8 11-515/9 11-515/7 11-515/10
ANSE11-515/11EA 277 EUR
ANSE11-515/11 ANSE11-515/6 ANSE11-515/8 ANSE11-515/9 ANSE11-515/7 ANSE11-515/10
Industrial gloves, HyFlex® 11-515
Handschoenen
This Kevlar®/stainless steel/Spandex glove with foam nitrile palm coating combines high visibility with superior cut resistance and durability.
  • Ability to handle sharp parts and knives with confidence, increasing worker output and security in light oil or dry applications
  • Provides ANSI Level 4 abrasion resistance for longer life
  • New coating technology provides increased wear in high stress areas
  • Silicone-free and latex-free for worker and product protection
  • Ergonomically designed so users are more likely to wear the glove
  • Helps keep workers visible in low light situations thus reducing the opportunities for hand injuries to occur
  • Extended cuff provides additional cut protection in the highly sensitive wrist area
The HyFlex® 11-515 glove has several outstanding features, including INTERCEPT TECHNOLOGY® yarn developed by Ansell and DuPont™, which incorporates Kevlar® Stretch Armor Technology.
Recommended for: Press shop operations; handling metal sheets & panels; inspection, selecting, checking parts; processing sheets, coils, tubes; cutting glasses, sheets, panels
Certificaten: EN 388:2016 (4X42E); Cat. II
Verpakking: 12 pairs per bag.