Potassium carbonate - sodium carbonate analytical reagent, Supelco®
Leverancier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
M-Clarity™ quality level = MQ100
1.06683.0500
1.06683.2500
1.06683.0500EA
90.9
EUR
1.06683.0500
1.06683.2500
Potassium carbonate - sodium carbonate analytical reagent, Supelco®
Potassium carbonate - sodium carbonate
Formule:
CKNaO₃ Dichtheid: 2,47 g/cm³ (20 °C) |
MDL Nummer:
MFCD00053407 CAS nummer: 10424-09-6 |
Specification Test Results
Solubility | (20 °C) soluble |
Bulk Density | 1750 kg/m3 |
Density | 2.47 g/cm3 (20 °C) |
pH | 11.5 (50 g/l, H2O, 20 °C) |