Aluminium carbide
Leverancier: Roth Carl
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3094.1
3094.2
ROTH3094.1EA
0
EUR
ROTH3094.1
ROTH3094.2
Aluminium carbide
Aluminium carbide
Formule:
C₃Al₄ Smeltpunt: 2100 °C Dichtheid: 2,36 g/cm³ (20 °C) |
MDL Nummer:
MFCD00003425 CAS nummer: 1299-86-1 EINECS: 215-076-2 UN: 1394 ADR: 4.3,II Merck Index: 12,00345 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...