Silicon dioxide for cleaning of platinum crucibles, calcinated, crude, Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide
18648-2.5KG 18648-50KG 18648-6X2.5KG
18648-2.5KGEA 85.3 EUR
18648-2.5KG 18648-50KG 18648-6X2.5KG
Silicon dioxide for cleaning of platinum crucibles, calcinated, crude, Fluka™
Silicon dioxide
Formule: SiO₂
MW: 60.08 g/mol
Kookpunt: 2230
Smeltpunt: 1719
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011232
CAS nummer: 14808-60-7
EINECS: 238-878-4
Merck Index: 14,08493