Chloorpyrifos
Leverancier: EHRENSTORFER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
C11600000 CR11600000
EHERC11600000EA 86.1 EUR
EHERC11600000 EHERCR11600000
Chloorpyrifos
Chloorpyrifos
Formule: C₉H₁₁Cl₃NO₃PS
Kookpunt: 276 °C (1013 hPa)
Smeltpunt: 41…42 °C
Dichtheid: 1,4 g/cm³ (20 °C)
CAS nummer: 2921-88-2
EINECS: 220-864-4
UN: 2811
ADR: 6.1,III
Merck Index: 12,02242
Order Now
Specificaties
Environmentally Preferable