Nickel(II) acetylacetonate for synthesis, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Bis(2,4-pentanedionato-O,O')nickel (II), Nickel (II) bis(2,4-pentanedionato-O,O'), Nickel acetylacetonate, Nickel (II) 2,4-pentanedionate
M-Clarity™ quality level = MQ200
8.06715.0025 8.06715.0100 8.06715.9025
8.06715.0025EA 46 EUR
8.06715.0025 8.06715.0100 8.06715.9025
Nickel(II) acetylacetonate for synthesis, Sigma-Aldrich®
Nickel(II) acetylacetonate
Formule: C₁₀H₁₄NiO₄
MW: 256.91 g/mol
Kookpunt: 220
Smeltpunt: 208
Dichtheid: 1.455 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000024
CAS nummer: 3264-82-2
EINECS: 221-875-7
Merck Index: 12,06584
Order Now
Specification Test Results
Solubility 4.8 g/l (20 °C)
Melting Point 208 °C
Molar Mass 256.91 g/mol
Bulk Density 470 kg/m3
Boiling Point 220 - 235 °C (14 hPa)
Vapor Pressure 2.7 hPa (110 °C)
Synonyms Bis(acetylacetonato)nickel(II), Bis(2,4-pentanedionato) nickel(II)
Hill Formula C10H14NiO4