Industrial gloves, ActivArmr® 43-216
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
43-216/11 43-216/9
ANSE43-216/11EA 94 EUR
ANSE43-216/11 ANSE43-216/9
Industrial gloves, ActivArmr® 43-216
Handschoenen
Heavy-duty welding glove for demanding welding and thermal job tasks.
  • Reliable protection for demanding welding and thermal job tasks
  • Suitable for stick and MIG welding, as well as torch cutting and grinding
  • Comfortable cotton lining provides thermal insulation and extended comfort
  • Long cuff extends protection over the forearm
ActivArmr® 43-216 has an improved 3-dimensional thumb design that greatly increases fine dexterity for precise welding control. Its premium split cowhide leather and welted seams with Kevlar® thread ensure durability in use while providing comfort and protection for the wearer.
High levels of durability, control, and protection from heat, flame, sparks, and puncture.
Certificaten: EN 388:2016 (3243B), EN407 (413X4X); Cat. III