Chemical resistant overalls, AlphaTec® 1800 Ts PLUS, Model 122
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WH18T-00122-02 WH18T-00122-03 WH18T-00122-04 WH18T-00122-05 WH18T-00122-06 WH18T-00122-07 WH18T-00122-08 WH18T-00122-09
ANSEWH18T-00122-02EA 564 EUR
ANSEWH18T-00122-02 ANSEWH18T-00122-03 ANSEWH18T-00122-04 ANSEWH18T-00122-05 ANSEWH18T-00122-06 ANSEWH18T-00122-07 ANSEWH18T-00122-08 ANSEWH18T-00122-09
Chemical resistant overalls, AlphaTec® 1800 Ts PLUS, Model 122
Wegwerpkleding
Lightweight, breathable microporous laminate technology, offering Type 4/5/6 protection.
  • Protects against low hazard liquid spray and fine particulates
  • Comfortable moisture vapor permeable fabric to help reduce the risk of heat stress
  • Optimized body fit, improving wearer comfort and safety
  • Anti-static, tested according to EN 1149-5 and AATCC 76
  • Low linting to help reduce the risk of cross contamination in critical areas
  • Silicone free, critical in spray painting applications
Coveralls feature a 3-piece hood, single zipper with re-sealable storm flap, finger loops, and chin strap. Tunnelled elasticated wrists and hood. Attached boots with anti-slip soles. Garments are manufactured from non-woven microporous polyethylene laminate. Stitched and taped seams.
Recommended for cleanroom cleaning and preparing, body painting including surface inspection, production line support and maintenance, plant and machine cleaning/maintenance, fibre glass resin application, and powder handling.
Certificaten: Type 4-B; Type 5-B, Type 6-B, EN 1073-2; EN 14126; EN 1149-5; Category III
Verpakking: Individually packaged; size S-XL 40 pieces/carton, size 2XL-5XL 30 piece/carton.