Chemical resistant overalls, AlphaTec® 5000, Model 125-G02
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR50T-12502G02 OR50T-12508G02 OR50T-12504G02 OR50T-12505G02 OR50T-12507G02 OR50T-12509G02 OR50T-12506G02 OR50T-12503G02
ANSEOR50T-12502G02EA 1290 EUR
ANSEOR50T-12502G02 ANSEOR50T-12508G02 ANSEOR50T-12504G02 ANSEOR50T-12505G02 ANSEOR50T-12507G02 ANSEOR50T-12509G02 ANSEOR50T-12506G02 ANSEOR50T-12503G02
Chemical resistant overalls, AlphaTec® 5000, Model 125-G02
Wegwerpkleding
Durable, highly visible, multi-layer chemical barrier technology offering Type 3/4/5 protection.
  • Outstanding permeation performance to organic and inorganic chemicals, biological hazards and chemical warfare agents
  • Exceptional barrier protection, providing >480 minutes breakthrough time against 14 of 15 chemicals listed in EN ISO 6529
  • Highly visible bright orange colour for improved worker safety
  • Welded and taped seams, offering the highest barrier to liquids and particulates
  • Multi-layer barrier material is lightweight, yet strong and durable
  • Anti-static, tested according to EN 1149-5
Designed to protect, typical innovative features include dual zip systems, double cuffs with knitted inner cuffs, 2-piece hood, integrated socks with boot overflaps, and attached barrier® Gloves. Elasticated hood, waist, and boot overflaps.
Recommended for use in contaminated environments, chemical handling, filling/blending/charging of raw materials, transferring liquids and solids, opening and draining pumps valves or lines, loading and unloading of proces equipment, raw material sample collection, concentration mixing and palletisation, Hazmat response, assembly and disassembly of large pipes, and unexpected leakages spills or other releases.
Certificaten: Type 3-B; Type 4-B; Type 5-B; EN 1073-2; EN 14126; EN 1149-5; Category III
Verpakking: Packaged 8 pieces/carton.