Chemical resistant overalls, AlphaTec® 1500 PLUS, Model 111
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WH15S-00111-08 WH15S-00111-09
ANSEWH15S-00111-08EA 128 EUR
ANSEWH15S-00111-08 ANSEWH15S-00111-09
Chemical resistant overalls, AlphaTec® 1500 PLUS, Model 111
Wegwerpkleding
Breathable, anti-static SMS material offering Type 5/6 protection.
  • Proven protection, filters 99,9% of particles >3 μm
  • Air and water vapour permeable fabric helps reduce the risk of heat stress
  • Optimised body fit, improving wearer comfort and safety
  • Silicone-free, critical in spray painting applications
  • Anti-static, tested according to EN 1149-5
Coveralls feature a 3-piece hood, 2-way front zipper with re-sealable storm flap, and finger loops. Elasticated hood, wrists, waist, and ankles. Red stitching for ease of identification. Garments areMore Product Information