Methylene blue
Leverancier: Serva
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Basic Blue 9, Methylthioninium chloride, Methylene blue chloride, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride, Swiss Blue
29198.01
29198.02
SERA29198.01EA
29
EUR
SERA29198.01
SERA29198.02
Methylene blue
Methylene blue
Formule:
C₁₆H₁₈ClN₃S Smeltpunt: 180 °C Dichtheid: 1,757 g/cm³ (22 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00150008 CAS nummer: 61-73-4 EINECS: 200-515-2 Merck Index: 12,06137 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...