2-Aminopyrazine
Leverancier: Cayman Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
10007093-5 10007093-10 10007093-25 10007093-50
CAYM10007093-5EA 38.6 EUR
CAYM10007093-5 CAYM10007093-10 CAYM10007093-25 CAYM10007093-50
2-Aminopyrazine
2-Aminopyrazine
Formule: C₄H₅N₃
Kookpunt: 203 °C (1013 hPa)
Smeltpunt: 117…120 °C
MDL Nummer: MFCD00006137
CAS nummer: 5049-61-6
EINECS: 225-748-7