Certified thermometers with DKD certificate
Leverancier: Amarell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
L25948-FL/DKD4 L25904-FL/DKD3
AMARL25948-FL/DKD4EA 299 EUR
AMARL25948-FL/DKD4 AMARL25904-FL/DKD3
Certified thermometers with DKD certificate
Thermometers Analoge thermometers
These precision thermometers, with enclosed scale, are suitable for government testing and/or official calibration.
  • Total immersion
  • Colourless capillary
  • Laboratory milk glass
Filling: red spirit.
Ø: 8,0 - 9,0 mm
Lengte: 420 mm
Leveringsinformatie: Geleverd met een DKD-certificaat.