V sampling spatulas with product retention, SteriWare®
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8115A-100 8115A-100S 8115A-150 8115A-150S 8115A-250 8115A-250S 8115N-100 8115N-100S 8115N-150 8115N-150S 8115N-250 8115N-250S
300-0534EA 171 EUR
300-0534 300-0535 300-0536 300-0537 300-0538 300-0539 300-0540 300-0541 300-0542 300-0543 300-0544 300-0545
V sampling spatulas with product retention, SteriWare®
Spatels
These V spatulas are ideal for goods in and general powder sampling. The angled tip design keeps the powder in the sampler which is perfect for almost all powders. These spatulas can also be used for harder more viscous materials.
  • Robust PS construction
  • Available in 3 volumes - 100, 150 and 250 mm blade lengths
  • Individually bagged and produced in a cleanroom
  • Full batch traceability
2 widths are available. The 10 mm is a narrow design ideal for sampling small amounts from bottles. The 20 mm is the standard design for sampling from sacks, bags, kegs and containers.
Certificaten: These sampling tools conform to all the latest standards including FDA 177.1640, EU 10/2011, EC 1935/2004 and are BSE/TSE free. Batch and irradiation certificates are available. Both EU and FDA compliant.