Labels for BBP33 printers, Type B-345, PermaSleeve® HT
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
142977 142979 142981 142983 142985 142987 142973
BRDY142977EA 1375.17 EUR
BRDY142977 BRDY142979 BRDY142981 BRDY142983 BRDY142985 BRDY142987 BRDY142973
Labels for BBP33 printers, Type B-345, PermaSleeve® HT
Etiketten
PermaSleeve® HT PVDF wire marker sleeve specifically designed for high temperature and low vacuum outgassing applications.
  • Fade-resistant and flame-retardant
  • Temperature resistance from –55 to 255 °C
Certificaten: Meets NASA SP-R-0022A and tested ASTM E162, ASTM E662, SAE-AMS-DTL-23053/18 (class 2) MIL-STD-202 - SAE-AS-81531.