Labels for BBP33 printers, type B-428
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
142880 142881 143085 133828 133884 133996 153390 152878 152879 153391 153392 153389 152877
BRDY142880EA 291.6 EUR
BRDY142880 BRDY142881 BRDY143085 BRDY133828 BRDY133884 BRDY133996 BRDY153390 BRDY152878 BRDY152879 BRDY153391 BRDY153392 BRDY153389 BRDY152877
Labels for BBP33 printers, type B-428
Etiketten
Metallised polyester (B-428) labels are designed for applications that use barcodes, alphanumerics, graphic symbols and logos and require nameplate-like quality.
  • Matte, light gray finish gives labels a rating and serial plate appearance
  • Service temperature from –40 to +120 °C
Labels are designed to withstand numerous solvents and variable temperatures when applied to various surfaces.
Certificaten: UL approved, CSA accepted.