Labels for thermal transfer printers, Type B-428, Brady
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
030108 030125 030133 030144 030147 030247
BRDY030108EA 573.05 EUR
BRDY030108 BRDY030125 BRDY030133 BRDY030144 BRDY030147 BRDY030247
Labels for thermal transfer printers, Type B-428, Brady
Etiketten
Printable, Metallised polyester labels for thermal transfer printers.
  • Fuel/oill resistance
  • High heat resistance
  • Low temperature resistance
  • Solvent/chemical resistance
Certificaten: This material is UL, CSA and AGA approved with its respective ribbon.