Labels for BBP33 printers, type B-7641, PermaSleeve® LSZH
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
140920 140921 140922 140923 140924 140925 140928 140929 140930 140931 140932 140933 140936
BRDY140920EA 412.4 EUR
BRDY140920 BRDY140921 BRDY140922 BRDY140923 BRDY140924 BRDY140925 BRDY140928 BRDY140929 BRDY140930 BRDY140931 BRDY140932 BRDY140933 BRDY140936
Labels for BBP33 printers, type B-7641, PermaSleeve® LSZH
Etiketten
PermaSleeve® LSZH Low Smoke Zero Halogen Polyolefin wire marking sleeves offer outstanding fire safety properties and minimal smoke emission.
  • Military/industry grade
  • Temperature resistance from –30 to 105 °C
  • 3:1 Shrink ratio double side printable
Certificaten: Meets SAE-AS-81531, MIL-STD-202 and tested ASTM E162, ASTM E662.