Labels for BBP33 printers, type B-484
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
133816 133824 133830 133892 133786 133787 153114
BRDY133816EA 124.87 EUR
BRDY133816 BRDY133824 BRDY133830 BRDY133892 BRDY133786 BRDY133787 BRDY153114
Labels for BBP33 printers, type B-484
Etiketten
Ultra-aggressive adhesive and pliable for conforming to angular surfaces (B-484) are general ID labels for lean 5S, bins and production areas.
  • Various sizes in several durable material types
  • Service temperature: –40 to +115 °C
Also ideal for marking backer board, DIN rail, relays, PLCs, switches, solenoids, drive motors and other components inside panels. All recommended materials are flexible and have high-tack adhesives for adhesion to wide variety of surfaces and components.
Certificaten: UL, RoHS certified; CSA approved.