Pre-printed ID tapes
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8221A-01 8221A-02 8221A-03 8221A-04
300-0561EA 98.7 EUR
300-0561 300-0562 300-0563 300-0564
Pre-printed ID tapes
Tape
ID tape helps to prevent handling errors by allowing containers, pallets, bags and so on to be easily labelled with highly visible messaging. Operators can quickly identify the status of a container to prevent it from being used in error.
  • Use with standard tape dispensers
  • Medium strength adhesion
ID tape is an ideal way of labelling large or small items. These tape rolls come pre-printed with 'Quarantine', 'Sampled', 'Released' or 'Rejected'.
Verpakking: 10 rolls per box.