Spill kits, acid, base and battery acid
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
241135 241138 241139
BRDY241135EA 144.85 EUR
BRDY241135 BRDY241138 BRDY241139
Spill kits, acid, base and battery acid
Absorptiematerialen en bescherming tegen morsen
These acid spill kit, base spill kit and battery acid spill kit safely remove acid, base and battery acid spills respectively.
  • Highly absorbing pads to wipe down equipment and remove spills
  • SOC's to prevent spills from spreading on the workfloor
  • Neutralising polymers to neutralise acids or bases
  • Gloves and glasses to protect employees
  • Waste disposal bag to contain all used equipment
These kits each contain a selection of solutions that enable users to combat small to medium spills.
Leveringsinformatie: Each spill kit is included with practical,More Product Information