Ribbons for label printer BP-4000/BP-4320 printers
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
236791 236785 236787 236789 236790 236783 236786
BRDY236791EA 303.81 EUR
BRDY236791 BRDY236785 BRDY236787 BRDY236789 BRDY236790 BRDY236783 BRDY236786
Ribbons for label printer BP-4000/BP-4320 printers
Printerlinten
Strong environmental resistance against smearing and chemicals.
  • Standard performance wax ribbon for paper and temporary labelling
  • Reliable, sharp-edge print quality to maximise bar code readability
  • Wax/resin ribbon formulation
R7960 series ribbons are printed on the appropriate Brady label, gives good solvent resistance, lower for alcohol, high print quality and excellent smear resistance.
Certificaten: RoHS compliant with Directive 2011/65/EU and 2015/863. Part does not contain an SVHC.