Labels for thermal transfer printers, type B-473
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
029908 029912
BRDY029908EA 154.58 EUR
BRDY029908 BRDY029912
Labels for thermal transfer printers, type B-473
Etiketten
Glossy, electrostatically dissipative polyester labels for printed circuit boards.
  • Fuels or oils resistant
  • High and low temperatures resistant
  • Solvents or chemicals resistant
Certificaten: UL recognised, CSA accepted, meets surface resistances within the recommended range for absorbent ESD packaging materials as defined by ANSI/ESD S541-2008.