Labels for BBP33 printers, type B-481
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
143047 143055 133831
BRDY143047EA 175.39 EUR
BRDY143047 BRDY143055 BRDY133831
Labels for BBP33 printers, type B-481
Etiketten
StainerBondz™ super chemical resistant polyester (B-481) is designed for use in the slide staining process and other laboratory ID which requires superior chemical resistance.
  • Performs well in the commonly used hematoxilin and eosin slide staining process
  • Excellent resistance to a wide variety of chemicals, oils and water
  • Service temperature of –80 to 130 °C
StainerBondz™ B-481 matte white polyester has a permanent acrylic adhesive and is thermal transfer printable with B30 series printers.