Labels for BBP33 printers, type B-489
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
142958 142969 143052 143062
BRDY142958EA 121.29 EUR
BRDY142958 BRDY142969 BRDY143052 BRDY143062
Labels for BBP33 printers, type B-489
Etiketten
Ultra aggressive polyester (B-489A) with permanent adhesive is ideal for labeling products and general ID in both commercial and industrial applications.
  • Matte white polyester material has a permanent rubber based adhesive and is thermal transfer printable
  • Service temperature of –40 to 120 °C
Best for application on smooth and highly textured indoor surfaces including metal and low surface energy plastics.
Certificaten: UL approved; CSA accepted.