Signs, floor signs, ToughStripe™, corner 'L' marks
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
104435 104433
116-0921EA 30.95 EUR
116-0921 116-0922
Signs, floor signs, ToughStripe™, corner 'L' marks
Pictogrammen
ToughStripe™ products are used for marking aisles, passageways and storage locations, and pointing out important safety equipment and egress paths.
  • Floor marking products are made with B-514 polyester
  • Recommended for indoor use and dry environment
  • Resistant to harsh chemicals
0,203 mm thickness and service temperature range of ⁻18 °C to 54 °C.
Certificaten: English XL Variable Incidence Tribometer per ASTM / ANSI F1679. ANSI A1264.2­2006 and OSHA 1910.22