Xyleen EMPARTA® ACS analytical reagent
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Dimethylbenzene
M-Clarity™ quality level = MQ200
1.08633.2500 1.08633.4000
1.08633.2500EA 186 EUR
1.08633.2500 1.08633.4000
Xyleen EMPARTA® ACS analytical reagent
Xyleen
Formule: C₆H₄(CH₃)₂
MW: 106.17 g/mol
Kookpunt: 140
Smeltpunt: –47 °C
Vlampunt: 26 °C
MDL Nummer: MFCD00077264
CAS nummer: 1330-20-7
UN: 1307
ADR: 3 III
Merck Index: 13,10136
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2deefc7b7b67c10e64003e243ff3216fad27cf7d&vid=9fa01ddafd4814d5f34266ae7933463a29d92d19&prid=web&clid=SAPDAM