Avantor® Hichrom HI-WAX MS, GC Columns
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HI25-25-025-15G5EA 583 EUR
HI25-25-025-15G5 HI25-25-025-30G5 HI25-25-025-60G5 HI25-53-100-30G5 HI25-18-018-20 HI25-18-018-40 HI25-18-018-60 HI25-18-020-10 HI25-18-020-20 HI25-18-020-40 HI25-18-020-50 HI25-18-020-60 HI25-18-040-10 HI25-18-040-20 HI25-18-040-60 HI25-20-010-10 HI25-20-010-60 HI25-20-020-20 HI25-20-020-50 HI25-20-100-10 HI25-20-100-20 HI25-20-100-40 HI25-20-100-50 HI25-25-010-30 HI25-25-010-60 HI25-25-025-105 HI25-25-025-50 HI25-25-050-105 HI25-25-050-50 HI25-25-050-60 HI25-25-100-105 HI25-25-100-15 HI25-25-100-50 HI25-32-010-10 HI25-32-010-105 HI25-32-010-50 HI25-32-010-60 HI25-32-025-10 HI25-32-025-105 HI25-32-050-105 HI25-32-050-15 HI25-32-050-50 HI25-32-050-60 HI25-32-100-10 HI25-32-100-105 HI25-32-100-15 HI25-32-100-60 HI25-32-150-10 HI25-32-150-105 HI25-32-150-15 HI25-32-150-50 HI25-32-150-60 HI25-32-300-10 HI25-32-300-105 HI25-32-300-15 HI25-32-300-30 HI25-32-300-50 HI25-53-010-15 HI25-53-010-30 HI25-53-025-15 HI25-53-025-60 HI25-53-050-30 HI25-53-100-10 HI25-53-100-60 HI25-53-100-75 HI25-53-150-15 HI25-53-200-30 HI25-53-200-60 HI25-53-250-10 HI25-53-250-50 HI25-05-005-2 HI25-05-005-2.5 HI25-05-005-5 HI25-10-020-10 HI25-10-020-15 HI25-10-020-5 HI25-10-040-10 HI25-10-040-5 HI25-15-015-10 HI25-15-015-15 HIGCHI2553100305IN HIGCHI25-20-040-25 HIGCHI25-32-100-25
Avantor® Hichrom HI-WAX MS, GC Columns
Kolommen voor chromatografie
Very popular PEG high polarity MS phase for a wide range of applications, including (for example): essential oils, flavours and fragrances also alkanols. HI-WAX MS is an MS grade low bleeding cross-bonded column.
  • Phase composition: Cross-bonded Polyethylene Glycol (PEG) - MS grade
  • Long lifetime
Max. temperature limits may change depending on the film thickness.
Order Now
Specificaties
Phase name HI-WAX MS
Environmentally Preferable
Phase composition Polyethylene Glycol (PEG) - low bleeding
Polarity Polar stationary phase
Crossbond Yes
Temperatuurlimiet 270 °C
USP code G14, G15, G16