Absorbent pads, SXT Oil Only
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
813732 813731
BRDY813732EA 114.15 EUR
BRDY813732 BRDY813731
Absorbent pads, SXT Oil Only
Absorptiematerialen en bescherming tegen morsen
These extra-tough oil only pads absorb oil and petroleum based liquids.
  • Durable coverstock provides extra strength and lasts longer
  • Low-linting - makes SXT ideal as a wipe down sorbent by eliminating loose fibres
  • Soft and pliable sorbent makes it easy to use
  • Perforated pads
These are ideal sorbents when durability or abrasion-resistance is required.